Learn More
Brand™ Filter Tips, PP, Colorless, Filter PE, BIO-CERT™ Liquid Handling Quality
Cleanroom quality for contamination-free analyses. Perfect fit for the Transferpette™ micropipette and many other pipettes. The tip system for all requirements.
Brand: Brand™ 732704
592.95 SEK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Description
Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses. The raw materials are free from DiHEMDA and Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. BRAND offers pipette tips for volumes from 0.1 ÎĽl to 10 ml and filter tips for volumes from 0.1 ÎĽl to 1250 ÎĽl. They are tested for BRAND pipettes and many pipette from other manufacturers.
- BRAND pipette tips and filter tips as well as the corresponding TipBoxes are autoclavable at 121 °C (2 bar) acc. DIN EN 285.
- Sterile tips are manufactured under certified BIO-CERT™ quality. They are sterilized with beta radiation according to ISO 11 137 and the AAMI Guidelines.
- Non-self-sealing filter tips from BRAND have a PE filter that is free from chemical additives.
- Permeability is controlled by the distinct combination of pore size and filter length, so that no aerosols can reach the pipette shaft.
- BIO-CERT™ Liquid Handling Quality: Free of human DNA, RNase, DNase, Pyrogens, PCR-Inhibitors and ATP
Specifications
0.5 to 10 ÎĽL | |
Non-sterile | |
Ideal for biological samples that contain detergents such as TRITON™ X-100, SDS, Tween | |
TipBox: 5 boxes, 96 each | |
Yes |
Filtered | |
Grey | |
Filter Tip | |
480 | |
Polyethylene |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
0000012653 | Certificate of Analysis | 23/05/2025 | 732704 |
Promotions
-
Multiple Manufacturers
Genomics Powerhouse
Power your pipeline from PCR to sequencing.Promo Code: 24117Expires: 2025-08-24
New Offer Save Up to 92%