Learn More
Masterflex™ Polyethylene Straight Connector, pack of 100
Designed for quick and easy assembly without tools
Marca: Masterflex™ 30626-55
Detalles adicionales : Weight : 0.80000kg
Descripción
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Especificaciones
Straight Connector | |
Not Autoclavable | |
3/16 in. | |
3/16 in. | |
Translucent |
Barb Connection | |
HDPE | |
3/16 in. | |
Ethylene Oxide |