Learn More
Masterflex™ Polyethylene Straight Connector, pack of 100
Designed for quick and easy assembly without tools
1350.00 SEK - 1430.00 SEK
Specifications
Product Type | Straight Connector |
---|---|
Description | Barb Connection |
Autoclavable | Not Autoclavable |
Material | HDPE |
Sterilization Method | Ethylene Oxide |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11709669
|
Masterflex™
30625-87 |
5/16 in. |
1350.00 SEK
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
15333568
|
Masterflex™
30626-50 |
5/32 in. |
1380.00 SEK
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
11799659
|
Masterflex™
30626-55 |
3/16 in. |
1430.00 SEK
Pack of 100 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
Description
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
Straight Connector | |
Not Autoclavable | |
Ethylene Oxide |
Barb Connection | |
HDPE | |
Translucent |