Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Ground Joint
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Ground Joint enables smaller containers to be supported on top the existing reservior.
Brand:  Poulten & Graf GmbH 101 09260
 View more versions of this product
Product Code. 10106400
  • 846.00 SEK / Each
This item is not returnable. View return policy
Description
Description
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of Polypropylene
  • Adapter with ground joint
  • Can be used for most chemicals and liquids
Specifications
Specifications
Adapters for OPTIFIX and POLYFIX Dispensers
White
Polypropylene
1
Ground joint
Autoclavable
Adapters for OPTIFIX and POLYFIX dispensers
OPTIFIX and POLYFIX dispensers
TS 60
Product Suggestions
Product Suggestions
Videos
Videos
  • Poulten Graf FORTUNA OPTIMAT 3
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Poulten Graf FORTUNA OPTIMAT 3
    SDS
    Documents
    Documents
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Ground Joint > ThreadSize: TS 60