Learn More
BRAND™ Polypropylene Conical Ground Joint Stopper
Brand: BRAND™ 144425
This item is not returnable.
View return policy
Description
Specifications
Conical bottom | |
NS 19/26 | |
Polypropylene | |
Square |
1/Ea. | |
Autoclavable | |
Ground Joint Stopper |
Product Content Correction
Product Title
BRAND™ Polypropylene Conical Ground Joint Stopper > Size: 19/26 NS
Spot an opportunity for improvement?Share a Content Correction