Learn More
VITLAB™ Polypropylene Conical Straight Connectors
Brand:  VITLAB™ 80877
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15242809
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Straight Connector
For 4.8 to 8.12mm tubing I.D.
Conical
Polypropylene
20 Pack