Learn More
Bochem™ Polypropylene Forceps
Brand: Bochem™ 1840
This item is not returnable.
View return policy
Specifications
Each | |
130 mm | |
Forceps |
Polypropylene | |
Forceps |
Product Content Correction
Product Title
Bochem™ Polypropylene Forceps > Length: 130mm
Spot an opportunity for improvement?Share a Content Correction