Learn More
WTW™ Portable Universal LED Filter Photometer
A unique multi-parameter function of photometry and electro-chemical pH measurement
Brand:  WTW™ 251200
34386.10 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10287892
  • 35600.00 SEK / Each
Only null left
Estimated Shipment: 11-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
With its unique multi-parameter function of photometry and electro-chemical pH measurement, pHotoFlex™ pH shows its strength with more complex tasks in environmental management, such as water framework guidelines, during process monitoring with changing sites and for fish hatcheries by determining NH3 and CO2: The energy-efficient LED optics with 6 wavelengths and a captive adapter for versatile applications with 16 and 28 cuvettes allows the measurement of the smallest concentrations with more than 180 programs for standard parameters and coloration. A list of favorites for the most commonly used tests, sample ID numbers and more than 3,000 measurements per battery set facilitate field measurements. The integrated pH function is a complete electro-chemical pH measurement of pH 0-16 with automatic buffer recognition (TEC/NIST) by using the comprehensive WTW selection of pH measuring chains. The temperature compensation automatically takes place in the admissible measuring ranges from -5 … 100 °C. The WTW MultiCal™ routine allows the automatic calibration with up to 3 calibration points. For field use, there is, for instance, the maintenance-free SenTix™ 41 is recommended, for precision measurements in the lab, the glass electrode SenTix™ 81 is frequently used. The optional PC software LSdata offers GLP-compliant data evaluation with data filter and allows the input of up to 100 user-defined programs, automatically calculate these as methods and save them to the pHotoFlex™. For the convenient use in the field, there is a case set available, including the pH probe SenTix™ 41 as a mobile lab with work surface and storage compartment as well as a lot of accessories, such as pH standards, tripod, pipettes and replacement batteries. For lab use, the photometer can be used in conjunction with a LabStation (optional) with site-supplied power and fitted with an external barcode reader.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Portable Universal Photometer
LED
1.5 V
LED
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Portable Universal LED Filter Photometer > Product Type: Portable Universal Photometer