missing translation for 'onlineSavingsMsg'
Learn More
Potassium Phosphate, Dibasic, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™ Product Code.: 15568104

Potassium Phosphate, Dibasic, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15568104 Shop All J.T.Baker Products
2.5 kg, Poly Bottle, 2.50kg
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.50kg
This item is not returnable. View return policy

Product Code. 15568104

Brand: Avantor J.T.Baker 325205

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7758-11-4
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphateShow MoreShow Less
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Physical Form Powder
Quantity 2.5 kg
Formula Weight 174.18
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.00kg
Chemical Name or Material Potassium Phosphate, Dibasic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Phosphate, Dibasic, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.