missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POTEH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | POTEH |
---|---|
Concentration | 0.28 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883335
|
Proteintech
24837-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873335
|
Proteintech
24837-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POTEH belongs to the POTE family and contains 7 ANK repeats.Specifications
POTEH | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6S545 | |
23784 | |
POTEH Fusion Protein Ag20275 | |
Primary | |
-20°C | |
POTEH |
0.28 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A26C3, ACTBL1, ANKRD26 like family C member 3, POTE 22, POTE22, POTEH | |
POTEH | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
POTEH Rabbit anti-Human, Polyclonal, Proteintech