Learn More
POU4F3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21509-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of non-syndromic sensorineural deafness autosomal domit type 15.Specifications
POU4F3 | |
Polyclonal | |
Unconjugated | |
POU4F3 | |
Brain 3C, Brn 3C, BRN3C, DFNA15, POU class 4 homeobox 3, POU4F3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18998, 5459 | |
-20°C | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15319, Q63955 | |
Pou4f3 | |
POU4F3 Fusion Protein Ag16004 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
POU4F3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction