Learn More
POU4F3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21509-1-AP-150UL
 View more versions of this product
Product Code. 16881095
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This gene encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of non-syndromic sensorineural deafness autosomal domit type 15.
Specifications
Specifications
POU4F3
Polyclonal
Unconjugated
POU4F3
Brain 3C, Brn 3C, BRN3C, DFNA15, POU class 4 homeobox 3, POU4F3
Rabbit
Antigen Affinity Chromatography
RUO
18998, 5459
-20°C
Liquid
Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15319, Q63955
Pou4f3
POU4F3 Fusion Protein Ag16004
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
POU4F3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated