Learn More
Lenz Laborglasinstrumente™ Powder Flask with Conical Ground Joint
Brand: Lenz Laborglasinstrumente™ 03043037
601.49 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Duran Glass | |
Rotary Evaporators | |
60 mm |
Powder Flask | |
100 mL |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Powder Flask with Conical Ground Joint > Capacity: 100mL
Spot an opportunity for improvement?Share a Content Correction