Learn More
Lenz Laborglasinstrumente™ Powder Flask with Conical Ground Joint
Brand:  Lenz Laborglasinstrumente™ 03043070
 View more versions of this product
Product Code. 11806104
  • 1330.00 SEK / Each
Only null left
Estimated Shipment: 04-08-2025
Specifications
Duran Glass
Rotary Evaporators
126 mm
Powder Flask
1000 mL
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Powder Flask with Conical Ground Joint > Capacity: 1000mL