missing translation for 'onlineSavingsMsg'
Learn More

PPAPDC2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen PPAPDC2
Concentration 0.4 mg/mL
Applications Immunofluorescence, Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16884745
View Documents
Proteintech
26234-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16874745
View Documents
Proteintech
26234-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PPAPDC2 is a phosphatase that dephosphorylates Presqualene diphosphate (PSDP) into presqualene monophosphate (PSMP), suggesting that it may have important role in innate immunity. PSDP is a bioactive lipid that rapidly remodels to PSMP upon cell activation. PPAPDC2 displays diphosphate phosphatase activity with a substrate preference for PSDP > FDP > phosphatidic acid. PPAPDC2 activity is independent of Mg2+ and has been identified in activated human neutrophil (PMN) extracts. It is widely expressed in human tissues. Recent studies shows PPAPDC2 is a functional isoprenoid diphosphate phosphatase.
Specifications

Specifications

PPAPDC2
Immunofluorescence, Immunoprecipitation, Immunocytochemistry
Unconjugated
Rabbit
Human
Q8IY26
403313
PPAPDC2 Fusion Protein Ag23856
Primary
-20°C
PLPP6
0.4 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
PPAPDC2, PSDP
PLPP6
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PPAPDC2 Rabbit anti-Human, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.