Learn More
PPID Mouse anti-Human, Unconjugated, Clone: 1A2B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67632-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16880366
  • 1656.73 SEK / 20µL
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
The PPID antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PPID. This antibody recognizes human, mouse, rat, pig antigen. The PPID antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PPID
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, ELISA
1A2B1
Unconjugated
20 μL
5481
IgG1