missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12590-2-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. This protein interacts with the proteinase inhibitor eglin c and is localized in the nucleus. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
PPIL2 | |
Polyclonal | |
Unconjugated | |
PPIL2 | |
CYC4, Cyclophilin 60, Cyp 60, CYP60, hCyP 60, PPIase, PPIL2, Rotamase PPIL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23759, 360746, 66053 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13356, Q9D787 | |
Ppil2 | |
PPIL2 Fusion Protein Ag3296 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPIL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction