Learn More
PPP1R3F Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | PPP1R3F |
---|---|
Concentration | 0.76 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896845
|
Proteintech
55386-1-AP-20UL |
20 ÎĽL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886845
|
Proteintech
55386-1-AP-150UL |
150 ÎĽL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycogen-targeting subunit for protein phosphatase 1 (PP1).Specifications
PPP1R3F | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q6ZSY5, Q9JIG4 | |
54646, 89801 | |
Peptide | |
Primary | |
-20°C | |
PPP1R3F |
0.76 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Hb2E, PPP1R3F | |
Ppp1r3f | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
PPP1R3F Rabbit anti-Human, Mouse, Polyclonal, Proteintech