missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2CA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13482-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit.Specifications
PPP2CA | |
Polyclonal | |
Unconjugated | |
PPP2CA | |
PP2A alpha, PP2Ac, PP2CA, PPP2CA, Replication protein C, RP C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19052, 24672, 5515 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P63331, P67775 | |
Ppp2ca | |
PPP2CA Fusion Protein Ag4294 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPP2CA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction