missing translation for 'onlineSavingsMsg'
Learn More

PPP3CB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1662.46 SEK - 4700.76 SEK

Specifications

Antigen PPP3CB
Concentration 0.22 mg/mL
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16896615
View Documents
Proteintech
55148-1-AP-20UL
20 ÎĽL
1662.46 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16886615
View Documents
Proteintech
55148-1-AP-150UL
150 ÎĽL
4700.76 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is specific to PPP3CB.

PPP3CB is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin.
Specifications

Specifications

PPP3CB
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Human, Mouse
P16298, P20651, P48453
19056, 24675, 5532
Peptide
Primary
-20°C
PPP3CB
0.22 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CALNA2, CALNB, CAM PRP catalytic subunit, CNA2, PPP3CB
Ppp3cb
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PPP3CB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.