Learn More
PPP3R1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13210-1-AP-20UL
This item is not returnable.
View return policy
Description
Protein phosphatase 3 (PPP3, formerly PP2B, Calcineurin), a serine/ threonine protein phosphatase, is a heterodimer composed of one catalytic subunit (PPP3C, Calcineurin A) and one regulatory subunit (PPP3R, Calcineurin B). PPP3R has two isoforms, PPP3R1 and PPP3R2. PPP3Rl is ubiquitously expressed in different tissues and is coded by the PPP3R1 gene which was mapped recently to human chromosome 2. PPP3Rl has four functional calcium-binding sites.Specifications
PPP3R1 | |
Polyclonal | |
Unconjugated | |
PPP3R1 | |
Calcineurin subunit B type 1, CALNB1, CNA2, CNB, CNB1, PPP3R1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5534 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P63098 | |
PPP3R1 | |
PPP3R1 Fusion Protein Ag3992 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
PPP3R1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction