missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15429-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the palmitoyl-protein thioesterase family. The encoded glycosylated lysosomal protein has palmitoyl-CoA hydrolase activity in vitro, but does not hydrolyze palmitate from cysteine residues in proteins. Three transcript variants encoding different isoforms have been described for this gene, with one of the isoforms being inactive.Specifications
PPT2 | |
Polyclonal | |
Unconjugated | |
PPT2 | |
C6orf8, G14, Lysosomal thioesterase PPT2, PPT 2, PPT2, S thioesterase G14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54397, 54398, 9374 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35448, O70489, Q9UMR5 | |
Ppt2 | |
PPT2 Fusion Protein Ag7671 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction