missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPWD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17106-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes the C-terminal of PPWD1.
Putative peptidylprolyl isomerase (PPIase). PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. May be involved in pre-mRNA splicing.Specifications
PPWD1 | |
Polyclonal | |
Unconjugated | |
PPWD1 | |
KIAA0073, PPWD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23398, 238831, 294711 | |
-20°C | |
Liquid |
Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CEC6, Q96BP3 | |
PPWD1 | |
PPWD1 Fusion Protein Ag10785 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPWD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction