missing translation for 'onlineSavingsMsg'
Learn More

PRAME Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen PRAME
Concentration 0.14 mg/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16888513
View Documents
Proteintech
11438-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16878513
View Documents
Proteintech
11438-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PRAME is a 509 amino acid protein belonging to the PRAME family consisting of 7 LRR (leucine-rich) repeats. It is a tumor antigen expressed in human melanomas and is recognized by cytolytic T lymphocytes. It′s overexpression frequently observed in human cancers confers growth or survival advantages by antagonizing retinoic acid receptor (RAR) signaling. PRAME serves as a candidate marker for monitoring childhood acute leukemia. It is expressed in testis and acute leukemias.
Specifications

Specifications

PRAME
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Human
P78395
23532
PRAME Fusion Protein Ag1906
Primary
-20°C
PRAME
0.14 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
MAPE, OIP 4, OIP4, Opa interacting protein 4, PRAME
PRAME
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PRAME Rabbit anti-Human, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.