missing translation for 'onlineSavingsMsg'
Learn More

PRAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen PRAP1
Concentration 0.34 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16879323
View Documents
Proteintech
11932-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16869323
View Documents
Proteintech
11932-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Proline rich acidic protein 1 (PRAP1) is also named as PRO1195 or UPA. It may play an important role in maintaining normal growth homeostasis in epithelial cells. Mitotic arrest deficient 1 (MAD1) is a key component in mitotic checkpoint signaling. Recent study identified PRAP1 as a novel MAD1 binding partner using yeast-two hybrid screening. PRAP1 has four isoforms with MW 16-20 kDa and 10 kDa.
Specifications

Specifications

PRAP1
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse
Q80XD8, Q96NZ9
118471, 22264
PRAP1 Fusion Protein Ag2526
Primary
-20°C
PRAP1
0.34 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
PRAP1, PRO1195, proline rich acidic protein 1, RP11 122K13.6, UPA
PRAP1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PRAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.