Learn More
PRDM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14126-1-AP-150UL
 View more versions of this product
Product Code. 16882624
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
PRDM5, a novel member of PR domain protein family, contains a PR-domain at the-NH2 terminus followed by 16 zinc finger motifs. It is found to have a role in cell differentiation and tumorigenesis. Reports suggest PRDM5 causes G2/M arrest and apoptosis upon infection of tumor cells and has been found to be silenced in breast, ovarian and lung cancers, suggesting a potential clinical significance.
Specifications
Specifications
PRDM5
Polyclonal
Unconjugated
PRDM5
PFM2, PR domain containing 5, PR domain containing protein 5, PRDM5
Rabbit
Antigen Affinity Chromatography
RUO
11107, 689788, 70779
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9CXE0, Q9NQX1
Prdm5
PRDM5 Fusion Protein Ag5277
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRDM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated