Läs mer
Prednisolone 17-Acetate, TRC
Produktkod. 30417405
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417405 100 mg
30427562 25 mg
30418166 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417405 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP703835100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52628-64-5
Molekylformel C23 H30 O6
Molekylvikt (g/mol) 402.48
Synonym Prednisolone 17-Acetate, (11β)-17-(Acetyloxy)-11,21-dihydroxypregna-1,4-diene-3,20-dione
IUPAC-namn [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl] acetate
LEDER CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO
Specifikationer
Kemiskt namn eller material Prednisolone 17-Acetate
Kvantitet 100 mg
InChI formel InChI=1S/C23H30O6/c1-13(25)29-23(19(28)12-24)9-7-17-16-5-4-14-10-15(26)6-8-21(14,2)20(16)18(27)11-22(17,23)3/h6,8,10,16-18,20,24,27H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1
Formel vikt 402.2042