CAS RN 52628-64-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
Prednisolone 17-Acetate, TRC
CAS: 52628-64-5 Molekylformel: C23 H30 O6 Molekylvikt (g/mol): 402.48 Synonym: Prednisolone 17-Acetate,(11β)-17-(Acetyloxy)-11,21-dihydroxypregna-1,4-diene-3,20-dione IUPAC-namn: [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl] acetate LEDER: CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO
2
Prednisolone 17-Acetate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.