Learn More
PRIM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10773-1-AP-150UL
 View more versions of this product
Product Code. 16887373
  • 4652.90 SEK / 150µL
Description
The replication of DNA in eukaryotic cells is carried out by a complex chromosomal replication apparatus, in which DNA polymerase alpha and primase are two key enzymatic components. Primase, which is a heterodimer of a small subunit and a large subunit, synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication. The protein encoded by this gene is the small, 49 kDa primase subunit.
Specifications
Specifications
PRIM1
Polyclonal
Unconjugated
PRIM1
DNA primase 49 kDa subunit, DNA primase small subunit, p49, PRIM1
Rabbit
Antigen Affinity Chromatography
RUO
19075, 246327, 5557
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P20664, P49642
Prim1
PRIM1 Fusion Protein Ag1124
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
PRIM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated