Learn More
PRKACA Mouse anti-Human, Unconjugated, Clone: 4G10D7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67491-1-IG-20UL
 View more versions of this product
Product Code. 16880146
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The PRKACA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRKACA. This antibody recognizes human, mouse, rat antigen. The PRKACA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PRKACA
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
4G10D7
Unconjugated
20 μL
5566
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRKACA Mouse anti-Human, Unconjugated, Clone: 4G10D7, Proteintech > 20 μL