Läs mer
PRKG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21646-1-AP-150UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Phosphorylates PPP1R12A.Specifikationer
| PRKG1 | |
| Polyclonal | |
| Unconjugated | |
| PRKG1 | |
| cGK 1, cGK1, CGKI, cGKI alpha, cGKI BETA, PGK, PKG, PRKG1, PRKG1B, PRKGR1A, PRKGR1B, Protein Kinase G1 alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19091, 54286, 5592 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P0C605, Q13976 | |
| Prkg1 | |
| PRKG1 Fusion Protein Ag16333 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |