Learn More
PRLR Mouse anti-Human, Unconjugated, Clone: 2E10G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67292-1-IG-150UL
Description
The PRLR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRLR. This antibody recognizes human, mouse antigen. The PRLR antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PRLR | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, Immunohistochemistry, ELISA | |
2E10G5 | |
Unconjugated | |
150 μL | |
5618 | |
IgG1 |