missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRODH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22980-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a mitochondrial proline dehydrogenase that catalyzes the first step in proline degradation. Defects in this gene are a cause of hyperprolinemia type 1 and possibly susceptibility to schizophrenia 4 (SCZD4). The gene is located on chromosome 22q11.21, a region which has also been associated with the contiguous gene deletion syndromes DiGeorge syndrome and CATCH22 syndrome.Specifications
PRODH | |
Polyclonal | |
Unconjugated | |
PRODH | |
HSPOX2, p53 induced gene 6 protein, PIG6, POX, POX2, PRODH, PRODH1, PRODH2, Proline oxidase, Proline oxidase 2, SCZD4, TP53I6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19125, 5625, 680409 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43272, Q9WU79 | |
Prodh, Prodh1 | |
PRODH Fusion Protein Ag18429 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRODH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction