Learn More
Profilin 1 Mouse anti-Human, Unconjugated, Clone: 3A12E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67390-1-IG-150UL
Description
The Profilin 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Profilin 1. This antibody recognizes human, mouse, rat antigen. The Profilin 1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Profilin 1 | |
Monoclonal | |
1000 ÎĽg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunofluorescence | |
3A12E8 | |
Unconjugated | |
150 ÎĽL | |
5216 | |
IgG2b |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.