missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Prostaglandin E2 Parameter Assay Kit
Prostaglandin E2 Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ SKGE004B
This item is not returnable.
View return policy
Description
The Prostaglandin E2 Parameter Assay Kit quantifies multi-species Prostaglandin E2/PGE2 in cell culture supernates (51 ul), serum (51 ul), edta plasma (51 ul), heparin plasma (51 ul), urine (51 ul).
Specifications
39.00 - 2500pg/mL | |
Biotin | |
ELISA Kit | |
PGE2 Levels in Various Samples | |
Multi-species | |
1 Kit | |
Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
41.4pg/mL | |
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules. | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine | |
6 x 96-well Plates | |
ELISA | |
Cell Culture Supernates (51uL),Serum (51uL),EDTA Plasma (51uL),Heparin Plasma (51uL),Urine (51uL) | |
Dinoprostone, PGE2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Prostaglandin E2 Parameter Assay Kit > Six 96-well plates
Spot an opportunity for improvement?Share a Content Correction