missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF40B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PRPF40B |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Produktcode | Marke | Quantity | Preis | Menge & VerfĂĽgbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & VerfĂĽgbarkeit | |||||
16896704
|
Proteintech
16929-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886704
|
Proteintech
16929-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Spezifikation
PRPF40B | |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q6NWY9, Q80W14 | |
25766, 363000, 54614 | |
PRPF40B Fusion Protein Ag10361 | |
Primary | |
-20°C | |
PRPF40B |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Huntingtin yeast partner C, HYPC, PRPF40B | |
Prpf40b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Name des Produkts
PRPF40B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech