missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11171-1-AP-150UL
This item is not returnable.
View return policy
Description
Pre-mRNA splicing occurs in 2 sequential transesterification steps. The protein encoded by this gene is a component of both U2- and U12-dependent spliceosomes, and found to be essential for the catalytic step II in pre-mRNA splicing process. It contains several WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp8 protein. This gene is a candidate gene for autosomal domit retinitis pigmentosa.Specifications
PRPF8 | |
Polyclonal | |
Unconjugated | |
PRPF8 | |
HPRP8, p220, PRP8, PRP8 homolog, PRPC8, PRPF8, RP13, Splicing factor Prp8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10594, 192159, 287530 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6P2Q9, Q99PV0 | |
Prpf8 | |
PRPF8 Fusion Protein Ag1652 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRPF8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction