missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPSAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16790-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Seems to play a negative regulatory role in 5-phosphoribose 1-diphosphate synthesis.Specifications
PRPSAP1 | |
Polyclonal | |
Unconjugated | |
PRPSAP1 | |
PAP39, PRPSAP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5635, 64390, 67763 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14558, Q63468, Q9D0M1 | |
PRPSAP1 | |
PRPSAP1 Fusion Protein Ag10227 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |