missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPSAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PRPSAP1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836534
|
Proteintech
16790-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826534
|
Proteintech
16790-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Seems to play a negative regulatory role in 5-phosphoribose 1-diphosphate synthesis.Specifications
PRPSAP1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q14558, Q63468, Q9D0M1 | |
5635, 64390, 67763 | |
PRPSAP1 Fusion Protein Ag10227 | |
Primary | |
-20°C | |
PRPSAP1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
PAP39, PRPSAP1 | |
PRPSAP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |