missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11618-1-AP-20UL
This item is not returnable.
View return policy
Description
PRRG2 encodes a protein that is a single-pass transmembrane protein containing an N-terminal gamma-carboxyglutamic acid (Gla) domain and tandem Pro/Leu-Pro-Xaa-Tyr (PY) motifs at its C-terminal end. The Gla domain is exposed on the cell surface while the PY motifs are cytoplasmic. The PY motifs of the encoded protein have been shown to interact with YAP1, a WW domain-containing protein. Therefore, it is thought that the encoded protein may be part of a signal transduction pathway. Two transcript variants encoding different isoforms have been found for this gene.Specifications
PRRG2 | |
Polyclonal | |
Unconjugated | |
PRRG2 | |
PRGP2, Proline rich Gla protein 2, PRRG2, TMG2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361570, 5639, 65116 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14669, Q8R182 | |
Prrg2 | |
PRRG2 Fusion Protein Ag2197 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRRG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction