missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRUNE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18537-1-AP-20UL
This item is not returnable.
View return policy
Description
Phosphodiesterase (PDE) that has higher activity toward cAMP than cGMP, as substrate. It plays a role in cell proliferation, is able to induce cell motility and acts as a negative regulator of NME1.Specifications
PRUNE | |
Polyclonal | |
Unconjugated | |
PRUNE1 | |
DRES 17, DRES17, hPrune, HTCD37, Protein prune homolog, PRUNE, prune homolog (Drosophila) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
229589, 310664, 58497 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AYG3, Q86TP1, Q8BIW1 | |
PRUNE1 | |
PRUNE Fusion Protein Ag13275 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRUNE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction