missing translation for 'onlineSavingsMsg'
Learn More

PSF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16247-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825824

  • 4721.67 SEK / 150µL
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The yeast heterotetrameric GINS complex is made up of Sld5, Psf1, Psf2, and Psf3. The formation of this complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts (Ueno et al., 2005 ). See GINS1 for additional information about the GINS complex.
Specifications

Specifications

PSF2
Polyclonal
Unconjugated
GINS2
DC5, GINS complex subunit 2, GINS2, HSPC037, Pfs2, PSF2
Rabbit
Antigen Affinity Chromatography
RUO
272551, 292058, 51659
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D600, Q9Y248
GINS2
PSF2 Fusion Protein Ag9180
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers