missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PSF2 |
---|---|
Concentration | 0.39 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16835824
|
Proteintech
16247-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16825824
|
Proteintech
16247-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The yeast heterotetrameric GINS complex is made up of Sld5, Psf1, Psf2, and Psf3. The formation of this complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts (Ueno et al., 2005 ). See GINS1 for additional information about the GINS complex.Specifications
PSF2 | |
Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q9D600, Q9Y248 | |
272551, 292058, 51659 | |
PSF2 Fusion Protein Ag9180 | |
Primary | |
-20°C | |
GINS2 |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DC5, GINS complex subunit 2, GINS2, HSPC037, Pfs2, PSF2 | |
GINS2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |