missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16247-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The yeast heterotetrameric GINS complex is made up of Sld5, Psf1, Psf2, and Psf3. The formation of this complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts (Ueno et al., 2005 ). See GINS1 for additional information about the GINS complex.Specifications
PSF2 | |
Polyclonal | |
Unconjugated | |
GINS2 | |
DC5, GINS complex subunit 2, GINS2, HSPC037, Pfs2, PSF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
272551, 292058, 51659 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D600, Q9Y248 | |
GINS2 | |
PSF2 Fusion Protein Ag9180 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |