missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSG4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PSG4 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887593
|
Proteintech
10902-2-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877593
|
Proteintech
10902-2-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a pregcy-specific glycoprotein (PSG), one of several encoded by a cluster of similar genes on chromosome 19. This gene is a member of the carcinoembryonic antigen (CEA) gene family and may play a role in regulation of the innate immune system. Several transcript variants encoding different isoforms have been found for this gene.Specifications
PSG4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q00888 | |
5672 | |
PSG4 Fusion Protein Ag1167 | |
Primary | |
-20°C | |
PSG4 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CGM4, CGM4, PSG9, PS beta G 4, PS beta G 9, PSBG 4, PSBG 9, PSG4, PSG9 | |
PSG4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PSG4 Rabbit anti-Human, Polyclonal, Proteintech