missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA1 Rabbit anti-Human, Mouse, Rat, Clone: 5X3A6, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | PSMA1 |
---|---|
Clone | 5X3A6 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331883
|
Bio-Techne
NBP3-16190-20UL |
20 ÎĽg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18313066
|
Bio-Techne
NBP3-16190-100UL |
100 ÎĽg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSMA1 Monoclonal antibody specifically detects PSMA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PSMA1 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuroscience, Ubiquitin Proteasome Pathway | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
A synthetic peptide corresponding to a sequence within amino acids 164-263 of human PSMA1 (P25786). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
5X3A6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
5682 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PSMA1 Rabbit anti-Human, Mouse, Rat, Clone: 5X3A6, Novus Biologicals™