missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PSMC2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883804
|
Proteintech
14905-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873804
|
Proteintech
14905-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May be involved in transcriptional regulation.Specifications
PSMC2 | |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P35998, P46471, Q63347 | |
19181, 25581, 5701 | |
PSMC2 Fusion Protein Ag6700 | |
Primary | |
-20°C | |
PSMC2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MSS1, Nbla10058, Protein MSS1, PSMC2, S7 | |
Psmc2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PSMC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech