missing translation for 'onlineSavingsMsg'
Learn More

PSMD10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12342-2-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890004

  • 1355.00 SEK / 20µL
Estimated Shipment: 23-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a subunit of the PA700/19S complex, which is the regulatory component of the 26S proteasome. The 26S proteosome complex is required for ubiquitin-dependent protein degradation. This protein is a non-ATPase subunit that may be involved in protein-protein interactions. Aberrant expression of this gene may paly a role in tumorigenesis. Two transcripts encoding different isoforms have been described. Pseudogenes have been identified on chromosomes 3 and 20.
Specifications

Specifications

PSMD10
Polyclonal
Unconjugated
PSMD10
Gankyrin, p28, p28(GANK), PSMD10
Rabbit
Antigen Affinity Chromatography
RUO
116722, 53380, 5716
-20°C
Liquid
Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75832, Q9Z2X2, Q9Z2X3
Psmd10
PSMD10 Fusion Protein Ag2994
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers