missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMD8 (Human) Recombinant Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
- Sequence: MYEQLKGEWNRKSPNLSKCGEELGRLKLVLLELNFLPTTGTKLTKQQLILARDILEIGAQWSILRKDIPSFERYMAQLKCYYFDYKEQLPESAYMHQLLGLNLLFLLSQNRVAEFHTELERLPAKDIQTNVYIKHPVSLEQYLMEGSYNKVFLAKGNIPAESYTFFIDILLDTIRDEIAGCIEKAYEKILFTEATRILFFNTPKKMTDYAKKRGWVLGPNNYYSFASQQQKPEDTTIPSTELAKQVIEYARQLEMIV
Specifications
Specifications
Accession Number | NP_002803 |
Gene ID (Entrez) | 5714 |
Name | proteasome (prosome, macropain) 26S subunit, non-ATPase, 8 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Quantity | 25 μg |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Gene Alias | HIP6, HYPF, MGC1660, Nin1p, Rpn12, S14, p31 |
Gene Symbol | PSMD8 |
Species | Wheat Germ (in vitro) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ PSMD8 (Human) Recombinant Protein >
Spot an opportunity for improvement?Share a Content Correction