Learn More
PSMD9 Mouse anti-Human, Unconjugated, Clone: 1H2G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67338-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The PSMD9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PSMD9. This antibody recognizes human, mouse, rat antigen. The PSMD9 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PSMD9 | |
Monoclonal | |
940 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1H2G1 | |
Unconjugated | |
150 μL | |
5715 | |
IgG1 |