Learn More
PSMD9 Mouse anti-Human, Unconjugated, Clone: 1H2G1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67338-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839915
  • 4721.67 SEK / 150µL
Estimated Shipment: 03-04-2024
to see stock.
Description
Description
The PSMD9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PSMD9. This antibody recognizes human, mouse, rat antigen. The PSMD9 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
PSMD9
Monoclonal
940 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
1H2G1
Unconjugated
150 μL
5715
IgG1